CD19 Monoclonal Antibody (J3-129), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD19 Monoclonal Antibody (J3-129), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (J3-129), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0197-41, 46-0197-42)