CD40 Monoclonal Antibody (1C10), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD40 Monoclonal Antibody (1C10), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD40 Monoclonal Antibody (1C10), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0401-80, 46-0401-82)