CD45 Monoclonal Antibody (HI30), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD45 Monoclonal Antibody (HI30), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (HI30), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0459-42, 46-0459-41)