CD54 (ICAM-1) Monoclonal Antibody (HA58), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD54 (ICAM-1) Monoclonal Antibody (HA58), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD54 (ICAM-1) Monoclonal Antibody (HA58), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0549-42, 46-0549-41)