CD258 (LIGHT) Monoclonal Antibody (7-3 (7)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD258 (LIGHT) Monoclonal Antibody (7-3 (7)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD258 (LIGHT) Monoclonal Antibody (7-3 (7)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2589-42)