F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs
F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4010-82, 46-4010-80)