eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate
Invitrogen™

eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
46-4317-8025 μg
46-4317-82100 μg
Número do catálogo 46-4317-80
Preço (BRL)
1.423,80
Each
Quantity:
25 μg
Preço (BRL)
1.423,80
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser

Emit

710 nm

Excite

488 nm

Reported Application

Flow Cytometric Analysis

For Research Use Only
Especificações
Concentration0.2 mg/mL
Label or DyePerCP-eFluor 710
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity25 μg
Target MoleculeBiotin
ConjugatePerCP-eFluor 710
FormLiquid
Product LineeBioscience
Unit SizeEach

Frequently asked questions (FAQs)

Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.