TCR V alpha 2 Monoclonal Antibody (B20.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TCR V alpha 2 Monoclonal Antibody (B20.1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5812-80)