KLRG1 Monoclonal Antibody (2F1), PerCP-eFluor™ 710, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (2F1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (2F1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5893-80, 46-5893-82)