CX3CR1 Monoclonal Antibody (2A9-1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CX3CR1 Monoclonal Antibody (2A9-1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CX3CR1 Monoclonal Antibody (2A9-1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-6099-42, 46-6099-41)