IL-10 Monoclonal Antibody (JES3-9D7), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-10 Monoclonal Antibody (JES3-9D7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-10 Monoclonal Antibody (JES3-9D7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7108-42, 46-7108-41)