IL-22 Monoclonal Antibody (IL22JOP), PerCP-eFluor™ 710, eBioscience™ - FAQs
IL-22 Monoclonal Antibody (IL22JOP), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IL-22 Monoclonal Antibody (IL22JOP), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-7222-82, 46-7222-80)