Survivin Monoclonal Antibody (STLALYV), PerCP-eFluor™ 710, eBioscience™ - FAQs
Survivin Monoclonal Antibody (STLALYV), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Survivin Monoclonal Antibody (STLALYV), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9176-41, 46-9176-42)