Galectin 9 Monoclonal Antibody (RG9-35), PerCP-eFluor™ 710, eBioscience™ - FAQs
Galectin 9 Monoclonal Antibody (RG9-35), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Galectin 9 Monoclonal Antibody (RG9-35), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9211-82, 46-9211-80)