IDO Monoclonal Antibody (mIDO-48), PerCP-eFluor™ 710, eBioscience™ - FAQs
IDO Monoclonal Antibody (mIDO-48), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IDO Monoclonal Antibody (mIDO-48), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9473-82, 46-9473-80)