AHR Monoclonal Antibody (FF3399), PerCP-eFluor™ 710, eBioscience™ - FAQs
AHR Monoclonal Antibody (FF3399), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for AHR Monoclonal Antibody (FF3399), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9854-41, 46-9854-42)