HLA-A2 Monoclonal Antibody (BB7.2), PerCP-eFluor™ 710, eBioscience™ - FAQs
HLA-A2 Monoclonal Antibody (BB7.2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for HLA-A2 Monoclonal Antibody (BB7.2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9876-42, 46-9876-41)