Gata-3 Monoclonal Antibody (TWAJ), PerCP-eFluor™ 710, eBioscience™ - FAQs
Gata-3 Monoclonal Antibody (TWAJ), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Gata-3 Monoclonal Antibody (TWAJ), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9966-41, 46-9966-42)