CD273 (B7-DC) Monoclonal Antibody (122), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD273 (B7-DC) Monoclonal Antibody (122), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD273 (B7-DC) Monoclonal Antibody (122), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9972-82, 46-9972-80)