TCR alpha/beta Monoclonal Antibody (IP26), PerCP-eFluor™ 710, eBioscience™ - FAQs
TCR alpha/beta Monoclonal Antibody (IP26), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TCR alpha/beta Monoclonal Antibody (IP26), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9986-41, 46-9986-42)