CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9987-42, 46-9987-41)