Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCHECK® HEV Antibody ELISA Kit, porcine

Catalog number: 4600020

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PrioCHECK® HEV Antibody ELISA is a reliable and fast diagnostic test for monitoring and surveillance of Hepatitis E virus in pig herds. The PrioCHECK® HEV Antibody ELISA is the only commercially available ELISA that uses HEV genotype 1 and 3 antigens. The use of genotype 3 antigens (the HEV genotype that has actually been demonstrated to infect pigs) significantly enhances the specificity and sensitivity of the test.

The PrioCHECK® HEV Antibody ELISA is a diagnostic kit based on ELISA technology. It follows a four-step protocol consisting of sample preparation, sample incubation, conjugate incubation, and detection. Serum or meat juice samples are incubated in an ELISA plate coated with the recombinant HEV antigens 1 and 3. Anti-HEV antibodies present in the sample bind to the antigen. Plates are then washed and an anti-swine antibody labeled with horse radish peroxidase (HRP) is used to detect antibodies bound to the HEV antigen. Color development using tetramethyl-benzidine (TMB) substrate measured optically at a wavelength of 450 nm shows the presence of antibodies directed against HEV.
For Veterinary Use Only.

Specifications

Protein Family: Other Proteins
Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Product Line: PrioCHECK®, Prionics™
Product Size: 4500 samples
Target (Kit Named): HEV Antibody
Target Organism Class: Hepatitis E Virus (HEV)
Target Species (Named): Porcine
Target Species (Validated): Porcine

Contents & storage

50 plates

Documents

Manuals & protocols