Methyl oleate, 99%, analytical standard for GC
Methyl oleate, 99%, analytical standard for GC
Methyl oleate, 99%, analytical standard for GC
Thermo Scientific Chemicals

Methyl oleate, 99%, analytical standard for GC

CAS: 112-62-9 | C19H36O2 | 296.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 460380010
Price (EUR)
32,65
Online exclusive
36,60
Save 3,95
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
32,65
Online exclusive
36,60
Save 3,95
Each
Chemical Identifiers
CAS112-62-9
IUPAC Namemethyl (9E)-octadec-9-enoate
Molecular FormulaC19H36O2
InChI KeyQYDYPVFESGNLHU-ZHACJKMWSA-N
SMILESCCCCCCCC\C=C\CCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only