Ribavirin, 98%
Ribavirin, 98%
Ribavirin, 98%
Thermo Scientific Chemicals

Ribavirin, 98%

Ribavirin, CAS# 36791-04-5, is an antiviral agent and a guanosine analog, which stops viral RNA synthesis and mRNA capping. | CAS: 36791-04-5 | C8H12N4O5 | 244.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460480250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS36791-04-5
IUPAC Name1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-1,2,4-triazole-3-carboxamide
Molecular FormulaC8H12N4O5
InChI KeyIWUCXVSUMQZMFG-AFCXAGJDSA-N
SMILESNC(=O)C1=NN(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Powder
HPLC>=97.5 %
Specific optical rotation-33° to -37° (20°C, 589 nm) (1 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ribavirin is a broad-spectrum antiviral nucleoside analog
  • It inhibits viral genome synthesis

Applications

  • Ribavirin is an antiviral compound
  • Its metabolite, ribavirin 5'-phosphate, inhibits inosine monophosphate dehydrogenase (IMP), which is involved in GTP biosynthesis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Keppeke, G.D.; et al. IMP/GTP balance modulates cytoophidium assembly and IMPDH activity. Cell Div. 2018, 15,13:5.
  2. Li, W.; Shen, F.; Weber, G.; Ribavirin and quercetin synergistically downregulate signal transduction and are cytotoxic in human ovarian carcinoma cells. Oncol Res. 1999, 11(5), 243-7.