3,5-Dibromoaniline, 98%
3,5-Dibromoaniline, 98%
3,5-Dibromoaniline, 98%
Thermo Scientific Chemicals

3,5-Dibromoaniline, 98%

CAS: 626-40-4 | C6H5Br2N | 250.92 g/mol
Have Questions?
Catalog NumberQuantity
4605600505 g
Catalog number 460560050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS626-40-4
IUPAC Name3,5-dibromoaniline
Molecular FormulaC6H5Br2N
InChI KeyRVNUUWJGSOHMRR-UHFFFAOYSA-N
SMILESNC1=CC(Br)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid or crystalline solid
Appearance (Color)Brown to black
Infrared spectrumConforms
Melting point51°C to 57°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only