5-Methoxypsoralen, 98%
5-Methoxypsoralen, 98%
5-Methoxypsoralen, 98%
Thermo Scientific Chemicals

5-Methoxypsoralen, 98%

CAS: 484-20-8 | C12H8O4 | 216.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number 460660250
Price (USD)
569.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
569.00
Each
Chemical Identifiers
CAS484-20-8
IUPAC Name4-methoxy-7H-furo[3,2-g]chromen-7-one
Molecular FormulaC12H8O4
InChI KeyBGEBZHIAGXMEMV-UHFFFAOYSA-N
SMILESCOC1=C2C=COC2=CC2=C1C=CC(=O)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point189°C to 193°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only