Diethyl iminodicarboxylate, 98%
Diethyl iminodicarboxylate, 98%
Diethyl iminodicarboxylate, 98%
Diethyl iminodicarboxylate, 98%
Thermo Scientific Chemicals

Diethyl iminodicarboxylate, 98%

CAS: 19617-44-8 | C6H11NO4 | 161.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 460770010
Price (EUR)
33,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
33,00
Each
Add to cart
Chemical Identifiers
CAS19617-44-8
IUPAC Nameethyl N-(ethoxycarbonyl)carbamate
Molecular FormulaC6H11NO4
InChI KeyPQVSTLUFSYVLTO-UHFFFAOYSA-N
SMILESCCOC(=O)NC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)White to off-white
Appearance (Form)Low melting solid
Infrared spectrumConforms
Melting point49°C to 54°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only