Thermo Scientific™

Ammonium hydroxide, for HPLC, 35% solution in water, Thermo Scientific™

100mL, Each
Thermo Scientific™

Ammonium hydroxide, for HPLC, 35% solution in water, Thermo Scientific™

100mL, Each
Quantity
Unit Size
Catalog number: 460801000
Price (USD)
40.40 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
1336-21-6
Titration with HCl
32 to 36 %
Appearance (Color)
Clear colorless
Lead (Pb)
=<0.05 ppm
Appearance (Form)
Solution
Iron (Fe)
=<0.2 ppm

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures