Tetracaine, 98%
Tetracaine, 98%
Tetracaine, 98%
Thermo Scientific Chemicals

Tetracaine, 98%

CAS: 94-24-6 | C15H24N2O2 | 264.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 460820250
Price (EUR)
112,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
112,00
Each
Chemical Identifiers
CAS94-24-6
IUPAC Name2-(dimethylamino)ethyl 4-(butylamino)benzoate
Molecular FormulaC15H24N2O2
InChI KeyGKCBAIGFKIBETG-UHFFFAOYSA-N
SMILESCCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point41°C to 46°C
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO497.5 to 102 % (on dried substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only