Imatinib mesylate, 98%
Imatinib mesylate, 98%
Imatinib mesylate, 98%
Thermo Scientific Chemicals

Imatinib mesylate, 98%

CAS: 220127-57-1 | C30H35N7O4S | 589.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 461080050
Price (USD)
640.65
Special offer
Online exclusive
Ends: 28-Dec-2026
753.00
Save 112.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
640.65
Special offer
Online exclusive
Ends: 28-Dec-2026
753.00
Save 112.35
Each
Chemical Identifiers
CAS220127-57-1
IUPAC NameN-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)-4-[(4-methylpiperazin-1-yl)methyl]benzamide; methanesulfonic acid
Molecular FormulaC30H35N7O4S
InChI KeyYLMAHDNUQAMNNX-UHFFFAOYSA-N
SMILESCS(O)(=O)=O.CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yelow or light beige
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
Water=<1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only