Levonorgestrel, 98%
Levonorgestrel, 98%
Levonorgestrel, 98%
Thermo Scientific Chemicals

Levonorgestrel, 98%

 
Have Questions?
產品號碼Quantity
4611000101g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 461100010
價格 (TWD)
-
Quantity:
1g
Request bulk or custom format
化學識別
CAS797-63-7
IUPAC Name(1R,3aS,3bR,9aR,9bS,11aS)-11a-ethyl-1-ethynyl-1-hydroxy-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular FormulaC21H28O2
InChI KeyWWYNJERNGUHSAO-XUDSTZEESA-N
SMILESCC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
檢視更多
規格Specification Sheet規格表
HPLC>=97.5 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Specific optical rotation-30° to -35° (20°C, 589 nm) (1 % in Chloroform)
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only