Texas Red™ Sulfonyl Chloride

Catalog number: 46115

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Thermo Scientific Texas Red Sulfonyl Chloride is a long-wavelength derivative of rhodamine that is modified with sulfonyl chloride for reaction to primary amines.

• Label antibodies for use as immunofluorescent probes
• Label oligonucleotides for hybridization probes
• Detect proteins in gels and on western blots

Properties of Rhodamine Dyes:
Thermo Scientific Pierce Rhodamine Dyes are mixtures of isomers with reactive groups attached at the 5- and 6-positions of the bottom ring. The properties of these isomers are indistinguishable in terms of excitation and emission spectra, and for protein applications there is no need to isolate a specific isomer.

Application Data:

Related Products
Pierce™ NHS-Rhodamine Antibody Labeling Kit
NHS-Rhodamine (5/6-carboxy-tetramethyl-rhodamine succinimidyl ester), mixed isomer
TRITC (5/6-tetramethyl-rhodamine isothiocyanate), mixed isomer
For Research Use Only. Not for use in diagnostic procedures.


Solubility: DMF (Dimethylformamide)
Label or Dye: Texas Red®
Product Size: 10 mg
Chemical Reactivity: Thiol
Excitation⁄Emission (nm): 596⁄615

Contents & storage

Storage: Upon receipt store desiccated at -20°C.


Manuals & protocols