Rifabutin, 98%, Thermo Scientific Chemicals
Rifabutin, 98%, Thermo Scientific Chemicals
Rifabutin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rifabutin, 98%, Thermo Scientific Chemicals

Rifabutin, CAS # 72559-06-09, is a broad-spectrum antibiotic with activity against mycobacteria, gram-positive and gram-negative bacteria.
Have Questions?
產品號碼Quantity
4612100101g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 461210010
價格 (TWD)
-
Quantity:
1g
Request bulk or custom format
化學識別
CAS72559-06-9
IUPAC Name(7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17-trihydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-1'-(2-methylpropyl)-6,23,32-trioxo-8,33-dioxa-24,27,29-triazaspiro[pentacyclo[23.6.1.1⁴,⁷.0⁵,³¹.0²⁶,³⁰]tritriacontane-28,4'-piperidin]-1,3,5(31),9,19,21,25,29-octaen-13-yl acetate
Molecular FormulaC46H62N4O11
InChI KeyATEBXHFBFRCZMA-VXTBVIBXSA-N
SMILESCO[C@H]1C=CO[C@@]2(C)OC3=C(C)C(O)=C4C(=O)C(NC(=O)C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)=C1NC5(CCN(CC(C)C)CC5)N=C1C4=C3C2=O
檢視更多
規格Specification Sheet規格表
Water=<2.5 %
HPLC>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)Red to violet
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Rifabutin is a compound with antibacterial activity most commonly used against Mycobacterium tuberculosis
Rifabutin is a derivative of Rifamycin S that acts through blocking RNA polymerase subunit beta (RpoB)
Rifabutin is an inducer as well as a substrate of CYP3A, a cytochrome P450 enzyme
Applications
Rifabutin may be used in pharmacokinetic laboratory studies and high-throughput drug screens
May be used in laboratory studies to determine drug resistance mutations present in Mycobacterium tuberculosis strains
RUO – Research Use Only
Literature references
Jamieson, F.B.; Guthrie, J.L.; Neemuchwala, A.; Lastovetska, O.; Melano, R.G.; Mehaffy, C. Profiling of rpoB mutations and MICs for rifampin and rifabutin in Mycobacterium tuberculosis. J Clin Microbiol. 2014, 52(6), 2157-62.
Cavusoglu, C.; Karaca-Derici, Y.; Bilgic, A. In-vitro activity of rifabutin against rifampicin-resistant Mycobacterium tuberculosis isolates with known rpoB mutations. Clin Microbiol Infect. 2004, 10(7), 662-5.
Doub, J.B.; Heil, E.L.; Ntem-Mensah, A.; Neeley, R.; Ching, P.R. Rifabutin Use in Staphylococcus Biofilm Infections: A Case Series. Antibiotics (Basel). 2020, 9(6), 326.
Sousa, M.; Pozniak, A.; Boffito, M. Pharmacokinetics and pharmacodynamics of drug interactions involving rifampicin, rifabutin and antimalarial drugs, Journal of Antimicrobial Chemotherapy, Volume 62, Issue 5, 2008, Pages 872–878.