This product ships via ground from US northeast.
Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals
Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals
Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4612500505g
4612500101g
46125025025g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461250050
Price (USD)
149.00
Each
Quantity:
5g
Request bulk or custom format
Price (USD)
149.00
Each
Chemical Identifiers
CAS202138-50-9
IUPAC Name(2E)-but-2-enedioic acid; bis({[(propan-2-yloxy)carbonyl]oxy}methyl) {[(2R)-1-(6-amino-9H-purin-9-yl)propan-2-yl]oxy}methanephosphonate
Molecular FormulaC23H34N5O14P
InChI KeyVCMJCVGFSROFHV-WZGZYPNHSA-N
SMILESOC(=O)C=CC(O)=O.CC(C)OC(=O)OCOP(=O)(CO[C@H](C)CN1C=NC2=C(N)N=CN=C12)OCOC(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Water=<1 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only