Product Image
Thermo Scientific Chemicals

Cedrol

222.37 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 461420250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77-53-2
IUPAC Name(1S,2R,5S,7R,8R)-2,6,6,8-tetramethyltricyclo[5.3.1.0¹,⁵]undecan-8-ol
Molecular FormulaC15H26O
InChI KeySVURIXNDRWRAFU-OGMFBOKVSA-N
SMILESC[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC[C@@]3(C)O
さらに表示
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Melting point80°C to 86°C
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only