Irinotecan hydrochloride trihydrate
Irinotecan hydrochloride trihydrate
Irinotecan hydrochloride trihydrate
Thermo Scientific Chemicals

Irinotecan hydrochloride trihydrate

CAS: 136572-09-3 | HCl·3H2O | 677.18 g/mol
製品番号(カタログ番号) 461540010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS136572-09-3
IUPAC Namehydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate trihydrate chloride
Molecular FormulaC33H45ClN4O9
InChI KeyKLEAIHJJLUAXIQ-JDRGBKBRSA-N
SMILES[H+].O.O.O.[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
さらに表示
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Appearance (Color)Yellow
Water7 to 9 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only