Rifaximin, 98%
Rifaximin, 98%
Rifaximin, 98%
Thermo Scientific Chemicals

Rifaximin, 98%

Have Questions?
Catalog NumberQuantity
4616800101g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461680010
Price (USD)
120.00
Each
Quantity:
1g
Request bulk or custom format
Price (USD)
120.00
Each
Chemical Identifiers
CAS80621-81-4
IUPAC Name(7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17,36-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22,30-octamethyl-6,23-dioxo-8,37-dioxa-24,27,33-triazahexacyclo[23.10.1.1⁴,⁷.0⁵,³⁵.0²⁶,³⁴.0²⁷,³²]heptatriaconta-1,3,5(35),9,19,21,25(36),26(34),28,30,32-undecaen-13-yl acetate
Molecular FormulaC43H51N3O11
InChI KeyNZCRJKRKKOLAOJ-XRCRFVBUSA-N
SMILESCO[C@H]1C=CO[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C1=C(N=C5C=C(C)C=CN15)C4=C3C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<5 %
Appearance (Color)Orange to red
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point200°C to 210°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only