Sodium tetrathionate dihydrate
Sodium tetrathionate dihydrate
Sodium tetrathionate dihydrate
Thermo Scientific Chemicals

Sodium tetrathionate dihydrate

CAS: 13721-29-4 | H4Na2O8S4 | 306.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 461920250
Price (USD)
148.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
148.00
Each
Chemical Identifiers
CAS13721-29-4
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange97.5 to 102.5 %
Solubility(1 g in 20 ml water) Clear to hazy colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only