Fluticasonpropionat, Thermo Scientific Chemicals
Fluticasonpropionat, Thermo Scientific Chemicals
Fluticasonpropionat, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fluticasonpropionat, Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
462101000100mg
462105000500mg
Katalognummer 462101000
Preis (EUR)
55,60
Each
Menge:
100mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
55,60
Each
Chemikalien-Kennzeichnungen
CAS80474-14-2
IUPAC Name(1R,2R,3aS,5S,9aS,9bR,10S,11aS)-5,9b-difluoro-1-{[(fluoromethyl)sulfanyl]carbonyl}-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl propanoate
Molecular FormulaC25H31F3O5S
InChI KeyWMWTYOKRWGGJOA-ZHLGSTKJSA-N
SMILESCCC(=O)O[C@@]1([C@H](C)C[C@H]2C3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Infrared spectrumConforms
HPLC>=96.0 %
Specific optical rotation+32° to +36° (20°C, 589 nm) (0.5% in CH2Cl2)
Appearance (Color)White to off-white
Appearance (Form)Powder
Mehr anzeigen
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only