Product Image
Thermo Scientific Chemicals

MOPSO sodium salt

CAS: 79803-73-9 | C7H14NNaO5S | 247.24 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 462140250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79803-73-9
IUPAC Namesodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate
Molecular FormulaC7H14NNaO5S
InChI KeyWSFQLUVWDKCYSW-UHFFFAOYNA-M
SMILES[Na+].OC(CN1CCOCC1)CS([O-])(=O)=O
さらに表示
Water=<0.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Total base>=98.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only