Raltegravir potassium salt
Raltegravir potassium salt
Raltegravir potassium salt
Thermo Scientific Chemicals

Raltegravir potassium salt

CAS: 871038-72-1 | C20H20FKN6O5 | 482.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462162500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS871038-72-1
IUPAC Namepotassium 4-{[(4-fluorophenyl)methyl]carbamoyl}-1-methyl-2-{2-[(5-methyl-1,3,4-oxadiazol-2-yl)formamido]propan-2-yl}-6-oxo-1,6-dihydropyrimidin-5-olate
Molecular FormulaC20H20FKN6O5
InChI KeyIFUKBHBISRAZTF-UHFFFAOYSA-M
SMILES[K+].CN1C(=O)C([O-])=C(N=C1C(C)(C)NC(=O)C1=NN=C(C)O1)C(=O)NCC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 %
HPLC>=97.5 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only