Salmeterol-Xinafoat
Salmeterol-Xinafoat
Salmeterol-Xinafoat
Thermo Scientific Chemicals

Salmeterol-Xinafoat

Have Questions?
KatalognummerMenge
462175000500mg
Katalognummer 462175000
Preis (EUR)
425,00
Each
Menge:
500mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
425,00
Each
Chemikalien-Kennzeichnungen
CAS94749-08-3
IUPAC Name1-hydroxynaphthalene-2-carboxylic acid; 4-(1-hydroxy-2-{[6-(4-phenylbutoxy)hexyl]amino}ethyl)-2-(hydroxymethyl)phenol
Molecular FormulaC36H45NO7
InChI KeyXTZNCVSCVHTPAI-UHFFFAOYNA-N
SMILESOC(=O)C1=CC=C2C=CC=CC2=C1O.OCC1=CC(=CC=C1O)C(O)CNCCCCCCOCCCCC1=CC=CC=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Water=<0.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only