Succinato de sumatriptán
Thermo Scientific Chemicals

Succinato de sumatriptán

CAS: 103628-48-4 | C18H27N3O6S | 413.49 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 462420050
Precio (EUR)
526,00
Each
Cantidad:
5 g
Pedido a granel o personalizado
Precio (EUR)
526,00
Each
Identificadores químicos
CAS103628-48-4
IUPAC Name1-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}-N-methylmethanesulfonamide; butanedioic acid
Molecular FormulaC18H27N3O6S
InChI KeyPORMUFZNYQJOEI-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O.CNS(=O)(=O)CC1=CC=C2NC=C(CCN(C)C)C2=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Water=<1 %
Infrared spectrumConforms
Melting point165°C to 169°C
HPLC>=97.5 %
Solubility(2.5 % in water) Clear colorless solution
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only