Product Image
Thermo Scientific Chemicals

Carvedilol

CAS: 72956-09-3 | C24H26N2O4 | 406.48 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 462450050
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS72956-09-3
IUPAC Name1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
Molecular FormulaC24H26N2O4
InChI KeyOGHNVEJMJSYVRP-UHFFFAOYNA-N
SMILESCOC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point115°C to 119°C
Loss on drying=<0.5 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only