Product Image
Thermo Scientific Chemicals

Valdecoxib

CAS: 181695-72-7 | C16H14N2O3S | 314.46 g/mol
Have Questions?
Catalog NumberQuantity
462522500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462522500
Price (EUR)
137,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
137,00
Each
Chemical Identifiers
CAS181695-72-7
IUPAC Name4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzene-1-sulfonamide
Molecular FormulaC16H14N2O3S
InChI KeyLNPDTQAFDNKSHK-UHFFFAOYSA-N
SMILESCC1=C(C(=NO1)C1=CC=CC=C1)C1=CC=C(C=C1)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point169°C to 173°C
Loss on drying=<0.5 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only