Lisinopril dihydrate
Lisinopril dihydrate
Lisinopril dihydrate
Thermo Scientific Chemicals

Lisinopril dihydrate

CAS: 83915-83-7 | C21H35N3O7 | 441.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462610010
Price (EUR)
64,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
64,50
Each
Chemical Identifiers
CAS83915-83-7
IUPAC Name(2S)-1-[(2S)-6-amino-2-{[(1S)-1-carboxy-3-phenylpropyl]amino}hexanoyl]pyrrolidine-2-carboxylic acid dihydrate
Molecular FormulaC21H35N3O7
InChI KeyCZRQXSDBMCMPNJ-ZUIPZQNBSA-N
SMILESO.O.NCCCC[C@H](N[C@@H](CCC1=CC=CC=C1)C(O)=O)C(=O)N1CCC[C@H]1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water8 to 9.5 %
Residue after ignition=<0.1 %
Infrared spectrumConforms
Solubility(1 % in water) Clear solution
Titration with NaOH98.0 to 102.0 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only