Product Image
Thermo Scientific Chemicals

Paroxetine hydrochloride hemihydrate, 98%

CAS: 110429-35-1 | C19H20FNO3 | 329.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462630050
Price (EUR)
243,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
243,00
Each
Chemical Identifiers
CAS110429-35-1
IUPAC Name(3S,4R)-3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine
Molecular FormulaC19H20FNO3
InChI KeyAHOUBRCZNHFOSL-YOEHRIQHSA-N
SMILESFC1=CC=C(C=C1)[C@@H]1CCNC[C@H]1COC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water2.0 to 3.0 %
Appearance (Color)White
Infrared spectrumConforms
Residue after ignition=<0.1 % (Platinum crucible)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only