Product Image
Thermo Scientific Chemicals

5-Fluoro-5'-deoxyuridine, Thermo Scientific Chemicals

CAS: 9-5-3094 | C9H11FN2O5 | 246.19 g/mol
Have Questions?
Catalog NumberQuantity
4627900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462790010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS3094-09-5
IUPAC Name1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11FN2O5
InChI KeyZWAOHEXOSAUJHY-ZIYNGMLESA-N
SMILESC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Melting point187°C to 193°C
HPLC>=97.5 %
Loss on drying=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only