Product Image
Thermo Scientific Chemicals

Ibandronate sodium salt monohydrate

CAS: 138926-19-9 | C9H24NNaO8P2 | 359.23 g/mol
Have Questions?
Catalog NumberQuantity
4631500101 g
Catalog number 463150010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS138926-19-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH96 to 104 % (on dry substance)
Loss on drying4.5 % to 8.5 %
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only