Product Image
Thermo Scientific Chemicals

Lacidipine

CAS: 103890-78-4 | C26H33NO6 | 455.55 g/mol
製品番号(カタログ番号)数量
4631800101 g
製品番号(カタログ番号) 463180010
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS103890-78-4
IUPAC Name3,5-diethyl 4-{2-[(1E)-3-(tert-butoxy)-3-oxoprop-1-en-1-yl]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular FormulaC26H33NO6
InChI KeyGKQPCPXONLDCMU-CCEZHUSRSA-N
SMILESCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1C=CC(=O)OC(C)(C)C)C(=O)OCC
さらに表示
Appearance (Form)Crystalline powder
Melting point177°C to 181°C
HPLC>=97.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only